- Organic syntheses based on 2?oxoglutaric acid. V. Syntheses of novel 2H?1, 4?benzothiazines and a 2, 5?dihydro?1, 5?benzothiazepine.
Organic syntheses based on 2?oxoglutaric acid. V. Syntheses of novel 2H?1, 4?benzothiazines and a 2, 5?dihydro?1, 5?benzothiazepine.
Journal of Heterocyclic Chemistry (1997)
Blitzke T, et al.