- Anion-catalyzed Phase-transfer Catalysis. II. Effects of Anionic Tetrakis [3, 5-bis (trifluoromethyl) phenyl] borate Catalyst in Phase-transfer-catalyzed Sulfonium Ylide Formation.
Anion-catalyzed Phase-transfer Catalysis. II. Effects of Anionic Tetrakis [3, 5-bis (trifluoromethyl) phenyl] borate Catalyst in Phase-transfer-catalyzed Sulfonium Ylide Formation.
Bulletin of the Chemical Society of Japan (1985)
Shiraki Y, et al.