Merck
  • Synthesis of (S)?and (R)?Harmicine from Proline: An Approach Toward Tetrahydro?β?carbolines.
Synthesis of (S)?and (R)?Harmicine from Proline: An Approach Toward Tetrahydro?β?carbolines.
European Journal of Organic Chemistry (2014)
Lood C S and Koskinen A M