Merck
  • Metabolomics Profiling of Serum and Urine in Three Beef Cattle Breeds Revealed Different Levels of Tolerance to Heat Stress.
Metabolomics Profiling of Serum and Urine in Three Beef Cattle Breeds Revealed Different Levels of Tolerance to Heat Stress.
Journal of agricultural and food chemistry (2018-06-16)
Yupeng Liao, Rui Hu, Zhisheng Wang, Quanhui Peng, Xianwen Dong, Xiangfei Zhang, Huawei Zou, Qijian Pu, Bai Xue, Lizhi Wang
ABSTRACT
This study was to determine differences in the global metabolic profiles of serum and urine of Xuanhan yellow cattle, Simmental crossbred cattle (Simmental × Xuanhan yellow cattle), and cattle-yaks (Jersey × Maiwa yak) under heat stress (temperature-humidity index remained above 80 for 1 week). A total of 55 different metabolites associated with the three breeds were identified in the serum and urine samples by gas chromatography-mass spectrometry. The metabolic adaptations to heat stress are heterogeneous. Cattle-yaks mobilize a greater amount of body protein to release glucogenic amino acids to supply energy, whereas the tricarboxylic acid cycle is inhibited. Simmental crossbred cattle mobilize a greater amount of body fat to use free fatty acids as an energy source. In comparison with Simmental crossbred cattle and cattle-yaks, Xuanhan yellow cattle have higher glycolytic activity and possess a stronger antioxidant defense system and are, in conclusion, more adapted to hot and humid environments.
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
D-Glutamine, ≥98% (HPLC)