Merck
  • [The expression of CD19 in 210 cases of childhood acute leukemia and its significance].
[The expression of CD19 in 210 cases of childhood acute leukemia and its significance].
Zhonghua er ke za zhi = Chinese journal of pediatrics (2004-05-18)
Ying-hu Chen, Yong-min Tang, Hong-qiang Shen, Hua Song, Shi-long Yang, Shu-wen Shi, Bai-qin Qian, Wei-qun Xu, Bo-tao Ning
ABSTRACT
To investigate the expression of CD19 on childhood acute leukemia (AL) and its significance, and to provide evidence for the diagnosis and differential diagnosis as well as monoclonal antibody-targeting treatment of leukemia. There were 210 cases of childhood AL, of which 130 cases were male and 80 were female with a mean age of 9 years old. Using a panel of 27 fluorochrome directly labeled monoclonal antibodies, 210 samples from the patients were analyzed with CD45/SSC double parameters and multi-color flow cytometry to determine the expression of CD19. In the 93 cases of B lineage acute lymphoblastic leukemia (ALL), the positive rate (98.9%, 92/93) of CD19 was significantly higher than that of the other B cell related antigens, such as CD10 (88.2%, 82/93, P = 0.003), CD20 (24.7%, 23/93, P = 0.001) and CD22 (60.2%, 56/93, P = 0.001). CD19 was expressed on all 8 cases of B/myeloid (My) hybrid acute leukemia (HAL) and 1 case of B/T HAL, but was not expressed on all 24 cases of T lineage leukemia and 5 cases of T/My HAL. In the 79 cases of acute myeloid leukemia (AML), only 5 (6.3%) cases expressed CD19. The positive rate (6.3%) of CD19 on AML was significantly lower than that on B lineage ALL (98.9%, P = 0.001). The percentage of CD19 positive cells in B/My HAL (41.6% - 88.7% with a mean of 73.8%) was significant higher than that in CD19(+)-AML (21.4% - 50.4% with a mean of 24.2%; Run Sum test, P = 0.0084). Of the 210 cases, 102 were B lineage related AL including B lineage ALL, B/My HAL and B/T HAL. In B lineage related AL, the sensitivity and the specificity of CD19 was 99.0% (101/102) and 95.4% (13/108) while the positive predictive and the negative predictive values to B lineage were 95.3% (101/106) and 99.0% (103/104), respectively. Using CD19(+) as a single reagent to diagnose B lineage, the false positive rate was 4.6% (5/108) and the false negative rate was 1.0% (1/102) with a general diagnosis index (GDI) of 94.4% [GDI = 1-(false positive rate + false negative rate)]. CD19 is continuously and stably expressed on all stages of B lineage differentiation. It is a reliable cell membrane marker for diagnosing B lineage ALL and an ideal target for antibody-targeting treatment of leukemia as well; the expression degree of CD19 can be used to distinguish B/My HAL from CD19(+)-AML; CD19 didn't express on normal myeloid cells but did on some AML cells. Therefore it can be used to detect the minimal residual disease.