- Biochemical analysis of trehalose and its metabolizing enzymes in wheat under abiotic stress conditions.
Biochemical analysis of trehalose and its metabolizing enzymes in wheat under abiotic stress conditions.
Plant Science (2005)
El-Bashiti T, et al.