Merck
  • Involvement of microRNA-23b in TNF-?-reduced BMSC osteogenic differentiation via targeting runx2.
Involvement of microRNA-23b in TNF-?-reduced BMSC osteogenic differentiation via targeting runx2.
Journal of Bone and Mineral Metabolism (2017)
Deng L, et al.