Merck
  • Evaluation of Acid Accelerators for an Off-Grid PEM Fuel Cell Power Station Utilizing Solid Sodium Borohydride for Hydrogen Storage
Evaluation of Acid Accelerators for an Off-Grid PEM Fuel Cell Power Station Utilizing Solid Sodium Borohydride for Hydrogen Storage
ECS Transactions (2016)
Hsu L, et al.