Merck
Global site-specific analysis of glycoprotein N-glycan processing.
Nature protocols (2018-05-05)
Liwei Cao, Jolene K Diedrich, Yuanhui Ma, Nianshuang Wang, Matthias Pauthner, Sung-Kyu Robin Park, Claire M Delahunty, Jason S McLellan, Dennis R Burton, John R Yates, James C Paulson
ABSTRACT
N-glycans contribute to the folding, stability and functions of the proteins they decorate. They are produced by transfer of the glycan precursor to the sequon Asn-X-Thr/Ser, followed by enzymatic trimming to a high-mannose-type core and sequential addition of monosaccharides to generate complex-type and hybrid glycans. This process, mediated by the concerted action of multiple enzymes, produces a mixture of related glycoforms at each glycosite, making analysis of glycosylation difficult. To address this analytical challenge, we developed a robust semiquantitative mass spectrometry (MS)-based method that determines the degree of glycan occupancy at each glycosite and the proportion of N-glycans processed from high-mannose type to complex type. It is applicable to virtually any glycoprotein, and a complete analysis can be conducted with 30 μg of protein. Here, we provide a detailed description of the method that includes procedures for (i) proteolytic digestion of glycoprotein(s) with specific and nonspecific proteases; (ii) denaturation of proteases by heating; (iii) sequential treatment of the glycopeptide mixture with two endoglycosidases, Endo H and PNGase F, to create unique mass signatures for the three glycosylation states; (iv) LC-MS/MS analysis; and (v) data analysis for identification and quantitation of peptides for the three glycosylation states. Full coverage of site-specific glycosylation of glycoproteins is achieved, with up to thousands of high-confidence spectra hits for each glycosite. The protocol can be performed by an experienced technician or student/postdoc with basic skills for proteomics experiments and takes ∼7 d to complete.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Calcium chloride dihydrate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
IgM from human serum, reagent grade, ~95% (HPLC), buffered aqueous solution
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Supelco
Discovery® Glycan SPE Tube, bed wt. 250 mg, volume 3 mL, pk of 54
Supelco
Discovery® Glycan SPE Tube, bed wt. 500 mg, volume 6 mL, pk of 30
Supelco
Discovery® Glycan SPE Tube, 5 g, volume 60 mL, pk of 16
Supelco
Discovery® Glycan SPE Bulk Packing, pkg of 50 g
Supelco
Discovery® Glycan SPE Tube, bed wt. 1 g, volume 12 mL, pk of 20
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%