Merck
  • Birth of Piglets After Transfer of Embryos Cryopreserved by Cytoskeletal Stabilization and Vitrification
Birth of Piglets After Transfer of Embryos Cryopreserved by Cytoskeletal Stabilization and Vitrification
Biology of Reproduction (2000)
Dobrinsky JR