Merck
  • A83-01 inhibits TGF-Β-induced upregulation of Wnt3 and epithelial to mesenchymal transition in HER2-overexpressing breast cancer cells.
A83-01 inhibits TGF-Β-induced upregulation of Wnt3 and epithelial to mesenchymal transition in HER2-overexpressing breast cancer cells.
Breast Cancer Research and Treatment (2017)
Wu Y, et al.