Merck
  • The Use of Endometrial Cancer Patient?Derived Organoid Culture for Drug Sensitivity Testing Is Feasible
The Use of Endometrial Cancer Patient?Derived Organoid Culture for Drug Sensitivity Testing Is Feasible
International Journal of Gynecological Cancer : Official Journal of the International Gynecological Cancer Society (2017)
Girda E, et al.