Merck
  • Oxidative and inert pyrolysis on-line coupled to gas chromatography with mass spectrometric detection: On the pyrolysis products of tobacco additives
Oxidative and inert pyrolysis on-line coupled to gas chromatography with mass spectrometric detection: On the pyrolysis products of tobacco additives
International Journal of Hygiene and Environmental Health (2016)
Paschke, Meike and Hutzler, Christoph and Henkler, Frank and Luch, Andreas