- Analysis of endogenous aldehydes in human urine by static headspace gas chromatography-mass spectrometry
Analysis of endogenous aldehydes in human urine by static headspace gas chromatography-mass spectrometry
Journal of Chromatography A (2016)
Serrano, Maria and Gallego, Mercedes and Silva, Manuel