Merck
  • Synthesis, characterization, and bulk crosslinking of polybutadiene-block-poly (2-vinyl pyridine)-block-poly (tert-butyl methacrylate) block terpolymers.
Synthesis, characterization, and bulk crosslinking of polybutadiene-block-poly (2-vinyl pyridine)-block-poly (tert-butyl methacrylate) block terpolymers.
Polymer (2010)
Schacher F, et al.