- Detection of Receptor Heteromerization Using In Situ Proximity Ligation Assay
Detection of Receptor Heteromerization Using In Situ Proximity Ligation Assay
Current Protocols in Pharmacology / Editorial Board, S.J. Enna (editor-in-chief) ... [Et al.] (2016)
Gomes I, et al.