Merck
  • Hemidesmus indicus (L.) R. Br. extract inhibits the early step of herpes simplex type 1 and type 2 replication.
Hemidesmus indicus (L.) R. Br. extract inhibits the early step of herpes simplex type 1 and type 2 replication.
The new microbiologica (2018-06-07)
Francesca Bonvicini, Mariacaterina Lianza, Manuela Mandrone, Ferruccio Poli, Giovanna Angela Gentilomi, Fabiana Antognoni
ABSTRACT
Herpes simplex virus types 1 (HSV-1) and 2 (HSV-2) cause several clinically relevant syndromes in both adults and neonates. Despite the availability of efficient anti-HSV agents, the search for new therapeutic approaches is highly encouraged due to the increasing drug resistance of virus strains. Medicinal plants represent a source of potential bioactive compounds. In this context we evaluated the anti-herpetic activity of Hemidesmus indicus (L.) R. Br., a plant widely used in traditional Indian medicine. The hydroalcoholic extract prepared from roots was characterized by NMR and HPLC analysis and assayed in vitro by CPE reduction and virus infectivity assays to define its anti-viral effect. The extract's mechanism of action was investigated by virucidal and time-of-addition assays and by in vitro α-glucosidase inhibitory assay. The extract exhibited a remarkable anti-herpetic activity at 100 mg/mL, at non-cytotoxic concentration, through multiple mechanisms: it reduced the infectivity of viral particles released from infected cells possibly through its anti-ER α-glucosidase inhibitory activity and it inhibited the beginning stage of HSV infection acting as a virucide agent and/or preventing virus attachment to the host cell surface.
Sigma-Aldrich
2-Hydroxy-4-methoxybenzoic acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Hydroxy-4-methoxybenzaldehyde, 98%
Pricing and availability is not currently available.