Merck
  • Analysis of carbohydrates in plants by high-performance anion-exchange chromatography coupled with electrospray mass spectrometry
Analysis of carbohydrates in plants by high-performance anion-exchange chromatography coupled with electrospray mass spectrometry
Journal of Chromatography A (2005)
Guignard C, et al.