- Mechanism of inhibition of DNA gyrase by analogues of nalidixic acid: the target of the drugs is DNA
Mechanism of inhibition of DNA gyrase by analogues of nalidixic acid: the target of the drugs is DNA
Proceedings of the National Academy of Sciences of the USA (1985)
Shen LL and Pernet AG