- The application of EDTA in drug delivery systems: doxorubicin liposomes loaded via NH4EDTA gradient
The application of EDTA in drug delivery systems: doxorubicin liposomes loaded via NH4EDTA gradient
International journal of nanomedicine (2014)
Song Y, et al.