Merck
  • Comparison of the efficacy of ethylenediaminetetraacetic acid and tetracycline hydrochloride as root conditioning agents: An in vitro study
Comparison of the efficacy of ethylenediaminetetraacetic acid and tetracycline hydrochloride as root conditioning agents: An in vitro study
Journal of the International Clinical Dental Research Organization (2012)
Nanda T, et al.