Merck
  • Histone deacetylase Rpd3 regulates olfactory projection neuron dendrite targeting via the transcription factor Prospero
Histone deacetylase Rpd3 regulates olfactory projection neuron dendrite targeting via the transcription factor Prospero
The Journal of Neuroscience (2010)
Tea JS, et al.