Merck
  • Histone deacetylase inhibitors improve the replication of oncolytic herpes simplex virus in breast cancer cells
Histone deacetylase inhibitors improve the replication of oncolytic herpes simplex virus in breast cancer cells
PLoS ONE (2014)
Cody JJ, et al.