Merck
  • A Large-Bandgap Conjugated Polymer for Versatile Photovoltaic Applications with High Performance.
A Large-Bandgap Conjugated Polymer for Versatile Photovoltaic Applications with High Performance.
Advanced materials (Deerfield Beach, Fla.) (2015-07-15)
Maojie Zhang, Xia Guo, Wei Ma, Harald Ade, Jianhui Hou
ABSTRACT
A new copolymer PM6 based on fluorothienyl-substituted benzodithiophene is synthesized and characterized. The inverted polymer solar cells based on PM6 exhibit excellent performance with Voc of 0.98 V and power conversion efficiency (PCE) of 9.2% for a thin-film thickness of 75 nm. Furthermore, the single-junction semitransparent device shows a high PCE of 5.7%.
Sigma-Aldrich
PBDB-T-2F
Pricing and availability is not currently available.