- Safety and efficacy of self-assembling bubble carriers stabilized with sodium dodecyl sulfate for oral delivery of therapeutic proteins
Safety and efficacy of self-assembling bubble carriers stabilized with sodium dodecyl sulfate for oral delivery of therapeutic proteins
Journal of Controlled Release : Official Journal of the Controlled Release Society (2017)
Lin P Y, et al.