Merck
  • Increased plasma thrombin potential is associated with stable coronary artery disease: An angiographically-controlled study
Increased plasma thrombin potential is associated with stable coronary artery disease: An angiographically-controlled study
Thrombosis Research (2017)
Tosi F, et al.