Merck
  • Mutations in lipoprotein lipase that block binding to the endothelial cell transporter GPIHBP1
Mutations in lipoprotein lipase that block binding to the endothelial cell transporter GPIHBP1
Proceedings of the National Academy of Sciences of the USA (2011)
Voss CV, et al.