Merck
  • Novel models for studying the blood-brain and blood-eye barriers in Drosophila
Novel models for studying the blood-brain and blood-eye barriers in Drosophila
The Blood-Brain and Other Neural Barriers (2011)
Nag S, et al.