Merck
  • Evolutionary conservation of vertebrate blood-brain barrier chemoprotective mechanisms in Drosophila
Evolutionary conservation of vertebrate blood-brain barrier chemoprotective mechanisms in Drosophila
The Journal of Neuroscience (2009)
Mayer F, et al.