Merck
  • Development of a sandwich format, amperometric screen-printed uric acid biosensor for urine analysis.
Development of a sandwich format, amperometric screen-printed uric acid biosensor for urine analysis.
Analytical biochemistry (2012)
Kanyong P, et al.