Merck
  • Effect of two barley beta-glucan concentrates on in vitro glycaemic impact and cooking quality of spaghetti
Effect of two barley beta-glucan concentrates on in vitro glycaemic impact and cooking quality of spaghetti
Food Sci. Technol. (2011)
Chillo, S and Ranawana, DV and Henry, CJK