Merck
  • Culture of rodent spermatogonial stem cells, male germline stem cells of the postnatal animal
Culture of rodent spermatogonial stem cells, male germline stem cells of the postnatal animal
Methods in Cell Biology (2008)
Kubota Hiroshi and Brinster Ralph L