Merck
  • Effects of an epinephrine infusion on eye temperature and heart rate variability in bull calves
Effects of an epinephrine infusion on eye temperature and heart rate variability in bull calves
Journal of Dairy Science (2010)
Stewart M, et al.