Merck
  • Mechanisms of topoisomerase I (TOP1) gene copy number increase in a stage III colorectal cancer patient cohort
Mechanisms of topoisomerase I (TOP1) gene copy number increase in a stage III colorectal cancer patient cohort
Testing (2013)
Smith DH, et al.