Merck
  • Multiresidue method for the determination of avermectin and moxidectin residues in the liver using HPLC with fluorescence detection.
Multiresidue method for the determination of avermectin and moxidectin residues in the liver using HPLC with fluorescence detection.
Analyst (1998)
Roudaut B.