Merck
  • Metabolic activity of brown,?beige,? and white adipose tissues in response to chronic adrenergic stimulation in male mice
Metabolic activity of brown,?beige,? and white adipose tissues in response to chronic adrenergic stimulation in male mice
American Journal of Physiology. Endocrinology and Metabolism (2016)
Labbe SM, et al.