Merck
  • Deoxyribonucleic acid polymerase from the extreme thermophile Thermus aquaticus.
Deoxyribonucleic acid polymerase from the extreme thermophile Thermus aquaticus.
Journal of bacteriology (1976-09-01)
A Chien, D B Edgar, J M Trela
PMID8432
ABSTRACT
A stable deoxyribonucleic acid (DNA) polymerase (EC 2.7.7.7) with a temperature optimum of 80 degrees C has been purified from the extreme thermophile Thermus aquaticus. The enzyme is free from phosphomonoesterase, phosphodiesterase and single-stranded exonuclease activities. Maximal activity of the enzyme requires all four deoxyribonucleotides and activated calf thymus DNA. An absolute requirement for divalent cation cofactor was satisfied by Mg2+ or to a lesser extent by Mn2+. Monovalent cations at concentrations as high as 0.1 M did not show a significant inhibitory effect. The pH optimum was 8.0 in tris(hydroxymethyl)aminomethane-hydrochloride buffer. The molecular weight of the enzyme was estimated by sucrose gradient centrifugation and gel filtrations on Sephadex G-100 to be approximately 63,000 to 68,000. The elevated temperature requirement, small size, and lack of nuclease activity distinguish this polymerase from the DNA polymerase of Escherichia coli.
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Pricing and availability is not currently available.