- The structure of the human gene encoding protein gene product 9.5 (PGP9.5), a neuron-specific ubiquitin C-terminal hydrolase.
The structure of the human gene encoding protein gene product 9.5 (PGP9.5), a neuron-specific ubiquitin C-terminal hydrolase.
The Biochemical journal (1990-06-01)
I N Day, L J Hinks, R J Thompson
PMID2163617
ABSTRACT
Database search using a bovine thymus ubiquitin C-terminal hydrolase sequence indicated 54% sequence identity with the abundant human neuron-specific protein gene product 9.5 (PGP9.5), which was then shown to possess the same activity [Wilkinson, Lee, Deshpande, Duerksen-Hughes, Boss & Pohl (1989) Science 246, 670-673]. A yeast counterpart of the enzyme is also known. The human PGP9.5 gene, described here, spans 10 kb, contains nine exons and displays 5' features some common to many genes and some common with neurofilament neuron-specific enolase and Thy-1-antigen gene 5' regions.
Sigma-Aldrich
Ubiquitin C-terminal hydrolase L1 (UCH-L1), 50 µg, From human cDNA, expressed in E. coli. Ubiquitin C-terminal hydrolases are a family of cysteine hydrolases that catalyze the hydrolysis of amides, esters & thioesters of the C-terminus of ubiquitin.
Pricing and availability is not currently available.