Merck
  • Scope and limitations of a DMF bio-alternative within Sonogashira cross-coupling and Cacchi-type annulation.
Scope and limitations of a DMF bio-alternative within Sonogashira cross-coupling and Cacchi-type annulation.
Beilstein journal of organic chemistry (2016-11-11)
Kirsty L Wilson, Alan R Kennedy, Jane Murray, Ben Greatrex, Craig Jamieson, Allan J B Watson
ABSTRACT
Pd-catalysed C-C bond formation is an essential tool within the pharmaceutical and agrochemical industries. Many of these reactions rely heavily on polar aprotic solvents; however, despite their utility, these solvents are incompatible with the drive towards more sustainable chemical synthesis. Herein, we describe the scope and limitations of an alternative to DMF derived from renewable sources (CyreneTM) in Sonogashira cross-coupling and Cacchi-type annulations.
Sigma-Aldrich
Cyrene, BioRenewable, DMF and NMP Substitute
Pricing and availability is not currently available.