Merck
  • Establishment and in vitro culture of porcine spermatogonial germ cells in low temperature culture conditions
Establishment and in vitro culture of porcine spermatogonial germ cells in low temperature culture conditions
Stem Cell Research (2013)
Lee WY, et al.