Merck
  • Statistical analysis of elicitation strategies for thiarubrine A production in hairy root cultures of Ambrosia artemisiifolia
Statistical analysis of elicitation strategies for thiarubrine A production in hairy root cultures of Ambrosia artemisiifolia
Journal of Biotechnology (2000)
Bhagwath SG and Hjortso MA