Merck
  • PTEN regulates RANKL- and osteopontin-stimulated signal transduction during osteoclast differentiation and cell motility.
PTEN regulates RANKL- and osteopontin-stimulated signal transduction during osteoclast differentiation and cell motility.
The Journal of biological chemistry (2002-12-04)
Toshifumi Sugatani, Ulises Alvarez, Keith A Hruska
ABSTRACT
PTEN (also known as MMAC-1 or TEP-1) is a frequently mutated tumor suppressor gene in human cancer. PTEN functions have been identified in the regulation of cell survival, growth, adhesion, migration, and invasiveness. Here, we characterize the diverse signaling networks modulated by PTEN in osteoclast precursors stimulated by RANKL and osteopontin (OPN). RANKL dose-dependently stimulated transient activation of Akt before activation of PTEN, consistent with a role for PTEN in decreasing Akt activity. PTEN overexpression blocked RANKL-activated Akt stimulated survival and osteopontin-stimulated cell migration while a dominant-negative PTEN increased the actions of RANKL and OPN. PTEN overexpression suppressed RANKL-mediated osteoclast differentiation and OPN-stimulated cell migration. The PTEN dominant-negative constitutively induced osteoclast differentiation and cell migration. Our data demonstrate multiple roles for PTEN in RANKL-induced osteoclast differentiation and OPN-stimulated cell migration in RAW 264.7 osteoclast precursors.
Sigma-Aldrich
Rho Activation Assay Kit, PP1/PP2A Toolbox for the selective in vitro dephosphorylation of proteins.
Pricing and availability is not currently available.