Merck
  • Characterisation of Norway lobster (<I>Nephrops norvegicus</I>) hyaluronidase and comparison with sheep and bovine testicular hyaluronidase.
Characterisation of Norway lobster (&lt;I&gt;Nephrops norvegicus&lt;/I&gt;) hyaluronidase and comparison with sheep and bovine testicular hyaluronidase.
Food Chemistry (1999)
Krishnapillai, A.M., et al.