Merck
  • Chemical-defined and albumin-free generation of human atrial and ventricular myocytes from human pluripotent stem cells
Chemical-defined and albumin-free generation of human atrial and ventricular myocytes from human pluripotent stem cells
Stem Cell Research (2017)
Pei F, et al.