Merck
  • Measurements of dissolved methylmercury in natural waters using diffusive gradients in thin film (DGT)
Measurements of dissolved methylmercury in natural waters using diffusive gradients in thin film (DGT)
Journal of Environmental Monitoring (2006)
Clarisse O and Hintelmann H