Merck
  • Relationship between the Structure of Ionic Liquid and Its Enrichment Ability To Trace Fungicides from an Environmental Water Sample.
Relationship between the Structure of Ionic Liquid and Its Enrichment Ability To Trace Fungicides from an Environmental Water Sample.
Journal of Agricultural and Food Chemistry (2018)
Yang J, et al.