Merck
  • Development of a rapid, simple and sensitive HPLC-FLD method for determination of rhodamine B in chili-containing products.
Development of a rapid, simple and sensitive HPLC-FLD method for determination of rhodamine B in chili-containing products.
Food Chemistry (2014)
Qi P, et al.