- Variations in catechol-O-methyltransferase gene interact with parenting to influence attention in early development
Variations in catechol-O-methyltransferase gene interact with parenting to influence attention in early development
Neuroscience (2009)
Voelker P, et al.