Merck
  • Expression of adenosine receptors and vegf during angiogenesis and its inhibition by pentoxifylline?a study using zebrafish model
Expression of adenosine receptors and vegf during angiogenesis and its inhibition by pentoxifylline?a study using zebrafish model
Biomedicine and Pharmacotherapy (2016)
Nathan J, et al.