Merck
  • Determination of 17 beta-estradiol in pharmaceutical preparation by UV spectrophotometry and high performance liquid chromatography methods
Determination of 17 beta-estradiol in pharmaceutical preparation by UV spectrophotometry and high performance liquid chromatography methods
Arabian Journal of Chemistry (2017)
Yilmaz B and Kadioglu Y